2-呋喃甲酰腈 ≥98.0% (by GC)

供应商: TCI
Warning

Synonyms: 2-氰基呋喃

C1648-25G C1648-5G
TCIAC1648-25GEA 0 CNY
TCIAC1648-25G TCIAC1648-5G
2-呋喃甲酰腈 ≥98.0% (by GC)
2-呋喃甲酰腈
分子式: C₅H₃NO
分子量: 93.08 g/mol
沸点: 148°C
密度: 1.085
闪点: 35°C
MDL编号: MFCD00003223
CAS号: 617-90-3
EINECS: 210-537-4
UN号: 1993
ADR: 3 III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9a1498559e5326d72b1c1ad93c2f52477e543f60&vid=9a1498559e5326d72b1c1ad93c2f52477e543f60&prid=web&clid=SAPDAM

现在订购

产品技术规格

Appearance Colorless to Light yellow to Light orange clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR