2-Bromo-3,5-dichloropyridine ≥98.0% (by GC)
供应商: TCI
|
Warning
|
B3734-25G
B3734-5G
TCIAB3734-25GEA
0
CNY
TCIAB3734-25G
TCIAB3734-5G
2-Bromo-3,5-dichloropyridine ≥98.0% (by GC)
2-Bromo-3,5-dichloropyridine
分子式:
C₅H₂BrCl₂N 分子量: 226.89 g/mol |
MDL编号:
MFCD00233990 CAS号: 14482-51-0 |
|
产品技术规格
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 41.0 to 45.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...