4-(三氟甲基)氯苄 ≥97.0% (by GC)

供应商: TCI
Danger

Synonyms: α′-氯-α,α,α-三氟对二甲苯

T2488-25G T2488-5G
TCIAT2488-25GEA 0 CNY
TCIAT2488-25G TCIAT2488-5G
4-(三氟甲基)氯苄 ≥97.0% (by GC)
4-(三氟甲基)氯苄
分子式: C₈H₆ClF₃
分子量: 194.58 g/mol
沸点: 68°C
溶点: 19°C
密度: 1.31
闪点: 74°C
MDL编号: MFCD00040772
CAS号: 939-99-1
UN号: 3265
ADR: 8 II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a7ec0e4861f046afb169f9ec521fb0a8757ab977&vid=a7ec0e4861f046afb169f9ec521fb0a8757ab977&prid=web&clid=SAPDAM

现在订购

产品技术规格

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 97.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR