4-氯硫代苯甲酰胺 ≥97.0% (by HPLC, total nitrogen)
供应商: TCI
|
Danger
|
Synonyms:
4-Chlorobenzene-1-carbothioamide
C2053-25G
C2053-5G
TCIAC2053-25GEA
0
CNY
TCIAC2053-25G
TCIAC2053-5G
4-氯硫代苯甲酰胺 ≥97.0% (by HPLC, total nitrogen)
4-氯硫代苯甲酰胺
分子式:
C₇H₆ClNS 分子量: 171.65 g/mol 沸点: 276 溶点: 134 |
MDL编号:
MFCD00040956 CAS号: 2521-24-6 UN号: 2811 ADR: 6.1 III |
|
产品技术规格
Appearance | Light yellow to Amber to Dark green powder to crystal |
Purity (HPLC) | min. 97.0 area% |
Purity (with Total Nitrogen) | min. 97.0 % |
Melting point | 127.0 to 131.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...