双(三氯甲基)碳酸酯 ≥98.0% (by titrimetric analysis)
供应商: TCI
|
Danger
|
Synonyms:
三光气, 固体光气
T1467-25G
T1467-250G
TCIAT1467-25GEA
0
CNY
TCIAT1467-25G
TCIAT1467-250G
双(三氯甲基)碳酸酯 ≥98.0% (by titrimetric analysis)
双(三氯甲基)碳酸酯
分子式:
C₃Cl₆O₃ 分子量: 296.75 g/mol 沸点: 206°C 存储条件: Refrigerator |
MDL编号:
MFCD00062848 CAS号: 32315-10-9 EINECS: 250-986-3 UN号: 2928 ADR: 6.1 II |
|
产品技术规格
Appearance | White to Orange to Green powder to lump |
Purity (Argentmetric Titration) | min. 98.0 % |
Melting point | 77.0 to 82.0 deg-C |
Solubility in Toluene | very faint turbidity |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...