三乙基胺盐酸盐 ≥98.0% (by titrimetric analysis)
供应商: TCI
|
Warning
|
Synonyms:
Triethylamine hydrochloride
T0425-25G
T0425-500G
TCIAT0425-25GEA
0
CNY
TCIAT0425-25G
TCIAT0425-500G
三乙基胺盐酸盐 ≥98.0% (by titrimetric analysis)
三乙基胺盐酸盐
分子式:
C₆H₁₆ClN 分子量: 137.65 g/mol |
MDL编号:
MFCD00012500 CAS号: 554-68-7 EINECS: 209-067-2 Merck Index: 14,09666 |
|
产品技术规格
Appearance | White to Almost white powder to crystal |
Purity (Argentmetric Titration) | min. 98.0 % (calcd.on anh.substance) |
Water | max. 3.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...