4-乙酰氧基苯甲醛 ≥98.0% (by GC)
供应商: TCI
|
Warning
|
Synonyms:
对乙酰氧基苯甲醛
A1467-25G
A1467-5G
TCIAA1467-25GEA
0
CNY
TCIAA1467-25G
TCIAA1467-5G
4-乙酰氧基苯甲醛 ≥98.0% (by GC)
4-乙酰氧基苯甲醛
分子式:
C₉H₈O₃ 沸点: 265°C |
MDL编号:
MFCD00003384 CAS号: 878-00-2 EINECS: 212-898-3 |
|
产品技术规格
Appearance | Colorless to Light yellow clear liquid |
Purity (GC) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...