3-溴-2-萘酚 ≥96.0% (by GC)

供应商: TCI
Warning

Synonyms: 3-Bromo-2-hydroxynaphthalene, 3-Bromonaphthalen-2-ol

B3127-1G B3127-5G
TCIAB3127-1GEA 0 CNY
TCIAB3127-1G TCIAB3127-5G
3-溴-2-萘酚 ≥96.0% (by GC)
3-溴-2-萘酚
分子式: C₁₀H₇BrO
MDL编号: MFCD10000951
CAS号: 30478-88-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3d1cbe0501554ec840b01347d8aa51f53795bc0f&vid=3d1cbe0501554ec840b01347d8aa51f53795bc0f&prid=web&clid=SAPDAM

现在订购

产品技术规格

Appearance White to Light yellow powder to crystal
Purity (GC) min. 96.0 %
Melting point 80.0 to 84.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR