2-氨基-4-溴酚 ≥98.0% (by GC)

供应商: TCI
Warning

Synonyms: 5-Bromo-2-hydroxyaniline

A2271-25G A2271-5G
TCIAA2271-25GEA 0 CNY
TCIAA2271-25G TCIAA2271-5G
2-氨基-4-溴酚 ≥98.0% (by GC)
2-氨基-4-溴酚
分子式: C₆H₆BrNO
分子量: 188.02 g/mol
溶点: 133°C
MDL编号: MFCD00235171
CAS号: 40925-68-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=29d518c7c89711a3ee23b49db3351da4bb12fa15&vid=29d518c7c89711a3ee23b49db3351da4bb12fa15&prid=web&clid=SAPDAM

现在订购

产品技术规格

Appearance White to Gray to Brown powder to crystal
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 97.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR