5-Bromo-2-fluorophenol ≥98.0% (by GC, titration analysis)
供应商: TCI
|
Warning
|
B3064-25G
B3064-5G
TCIAB3064-25GEA
0
CNY
TCIAB3064-25G
TCIAB3064-5G
5-Bromo-2-fluorophenol ≥98.0% (by GC, titration analysis)
5-Bromo-2-fluorophenol
分子式:
C₆H₄BrFO 分子量: 191 g/mol 沸点: 54°C |
MDL编号:
MFCD07782066 CAS号: 112204-58-7 |
|
产品技术规格
Appearance | Colorless to Light yellow to Light orange clear liquid |
Purity (GC) | min. 98.0 % |
Purity (Neutralization titration) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...