1-Bromo-2-fluoro-4-nitrobenzene ≥95.0% (by GC)
供应商: TCI
|
Warning
|
Synonyms:
4-Bromo-3-fluoronitrobenzene
B3060-25G
B3060-5G
TCIAB3060-25GEA
0
CNY
TCIAB3060-25G
TCIAB3060-5G
1-Bromo-2-fluoro-4-nitrobenzene ≥95.0% (by GC)
1-Bromo-2-fluoro-4-nitrobenzene
分子式:
C₆H₃BrFNO₂ 分子量: 220 g/mol |
MDL编号:
MFCD05865088 CAS号: 185331-69-5 |
|
产品技术规格
Appearance | White to Orange to Green powder to crystal |
Purity (GC) | min. 95.0 % |
Melting point | 88.0 to 94.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...