4-溴-2,6-二氟苯甲醛 ≥98.0% (by GC)

供应商: TCI
Warning

B3446-1G B3446-100MG
TCIAB3446-1GEA 0 CNY
TCIAB3446-1G TCIAB3446-100MG
4-溴-2,6-二氟苯甲醛 ≥98.0% (by GC)
4-溴-2,6-二氟苯甲醛
分子式: C₇H₃BrF₂O
MDL编号: MFCD03094459
CAS号: 537013-51-7

现在订购

产品技术规格

Appearance White to Light yellow to Light orange powder to crystal
Purity (GC) min. 98.0 %
Melting point 75.0 to 79.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR