(5-Bromopentyl)benzene ≥98.0% (by GC)

供应商: TCI

Synonyms: 1-Bromo-5-phenylpentane

P1323-25G P1323-5G
TCIAP1323-25GEA 0 CNY
TCIAP1323-25G TCIAP1323-5G
(5-Bromopentyl)benzene ≥98.0% (by GC)
(5-Bromopentyl)benzene
分子式: C₁₁H₁₅Br
分子量: 227.14 g/mol
沸点: 148°C
MDL编号: MFCD01075177
CAS号: 14469-83-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a16edfc610d4f5596eb36c3dec41b08dfea10deb&vid=a16edfc610d4f5596eb36c3dec41b08dfea10deb&prid=web&clid=SAPDAM

现在订购

产品技术规格

Appearance Colorless to Light yellow to Light orange clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR