1-溴-6-氯己烷 ≥95.0% (by GC)
供应商: TCI
|
Warning
|
B1998-25G
B1998-500G
TCIAB1998-25GEA
0
CNY
TCIAB1998-25G
TCIAB1998-500G
1-溴-6-氯己烷 ≥95.0% (by GC)
1-溴-6-氯己烷
分子式:
BrH₂C(CH₂)₄CH₂Cl 分子量: 199.52 g/mol 沸点: 96°C 闪点: 117°C |
MDL编号:
MFCD00001019 CAS号: 6294-17-3 EINECS: 228-555-6 |
|
产品技术规格
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 95.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...