1-溴十三烷 ≥98.0% (by GC)

供应商: TCI
Warning

Synonyms: 十三烷基溴

B0935-25ML B0935-5ML
TCIAB0935-25MLEA 0 CNY
TCIAB0935-25ML TCIAB0935-5ML
1-溴十三烷 ≥98.0% (by GC)
1-溴十三烷
分子式: BrH₂C(CH₂)₁₁CH₃
分子量: 263.26 g/mol
沸点: 292
溶点: 6
存储条件: Ambient
MDL编号: MFCD00000227
CAS号: 765-09-3
EINECS: 212-138-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2c41469044b0a954f93405e762ec47606e421f05&vid=f544e04685c023a25d0e9add243286daf9f753d1&prid=web&clid=SAPDAM

现在订购

产品技术规格

Appearance Colorless to Light orange to Yellow clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR