1-溴辛烷 ≥98.0% (by GC)
供应商: TCI
|
Warning
|
Synonyms:
辛基溴
B0626-25ML
B0626-500ML
TCIAB0626-25MLEA
0
CNY
TCIAB0626-25ML
TCIAB0626-500ML
1-溴辛烷 ≥98.0% (by GC)
1-溴辛烷
分子式:
BrH₂C(CH₂)₆CH₃ 沸点: 201°C 溶点: –55 °C 密度: 1,112 g/cm³ (20 °C) 闪点: 92°C 存储条件: Freezer |
MDL编号:
MFCD00000276 CAS号: 111-83-1 EINECS: 203-912-9 UN号: 3082 ADR: 9,III Merck Index: 14,06764 |
|
产品技术规格
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 98.0 % |