1-溴壬烷 ≥98.0% (by GC)

供应商: TCI

Synonyms: 壬基溴

B0933-25ML B0933-500ML
TCIAB0933-25MLEA 0 CNY
TCIAB0933-25ML TCIAB0933-500ML
1-溴壬烷 ≥98.0% (by GC)
1-溴壬烷
分子式: BrH₂C(CH₂)₇CH₃
分子量: 207.15 g/mol
沸点: 201
溶点: -29
闪点: 90°C
存储条件: Ambient
MDL编号: MFCD00000278
CAS号: 693-58-3
EINECS: 211-755-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=25c0fdf7d320a209433e6ce82b5d592485a80494&vid=807dbbad41a8b4bc0d07e380a177a58a49d87752&prid=web&clid=SAPDAM

现在订购

产品技术规格

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR