1-溴庚烷 ≥98.0% (by GC)
供应商: TCI
|
Warning
|
Synonyms:
1-溴代庚烷
B0598-25G
B0598-500G
TCIAB0598-25GEA
0
CNY
TCIAB0598-25G
TCIAB0598-500G
1-溴庚烷 ≥98.0% (by GC)
1-溴庚烷
分子式:
BrH₂C(CH₂)₅CH₃ 沸点: 179 °C (1013 hPa) 溶点: –58 °C 密度: 1,1602 g/cm³ (20 °C) 闪点: 60°C 存储条件: Ambient |
MDL编号:
MFCD00000273 CAS号: 629-04-9 EINECS: 211-068-8 UN号: 1993 ADR: 3,III |
|
产品技术规格
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...