1-溴十七烷 ≥98.0% (by GC)
供应商: TCI
Synonyms:
十七基溴
B0937-25G
B0937-5G
TCIAB0937-25GEA
0
CNY
TCIAB0937-25G
TCIAB0937-5G
1-溴十七烷 ≥98.0% (by GC)
1-溴十七烷
分子式:
BrH₂C(CH₂)₁₅CH₃ 沸点: 349 °C (1013 hPa) 溶点: 32 °C 密度: 0,99 g/cm³ (20 °C) 存储条件: Ambient |
MDL编号:
MFCD00017891 CAS号: 3508-00-7 EINECS: 222-506-2 |
|
产品技术规格
Appearance | White or Colorless to Light yellow powder to lump to clear liquid |
Purity (GC) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...