1-溴癸烷 ≥98.0% (by GC)
供应商: TCI
|
Warning
|
Synonyms:
癸基溴
B0583-25ML
B0583-500ML
TCIAB0583-25MLEA
0
CNY
TCIAB0583-25ML
TCIAB0583-500ML
1-溴癸烷 ≥98.0% (by GC)
1-溴癸烷
分子式:
BrH₂C(CH₂)₈CH₃ 分子量: 221.18 g/mol 沸点: 139°C 溶点: –30°C 闪点: 94°C 存储条件: Ambient |
MDL编号:
MFCD00000221 CAS号: 112-29-8 EINECS: 203-955-3 |
|
产品技术规格
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...