1-Bromobutane ≥98.0% (by GC)
供应商: TCI
|
Danger
|
Synonyms:
Butyl bromide
B0560-25G
B0560-500G
TCIAB0560-25GEA
0
CNY
TCIAB0560-25G
TCIAB0560-500G
1-Bromobutane ≥98.0% (by GC)
1-Bromobutane
分子式:
BrH₂C(CH₂)₂CH₃ 分子量: 137.02 g/mol 沸点: 101°C 溶点: –112°C 闪点: 13°C 存储条件: Ambient |
MDL编号:
MFCD00000260 CAS号: 109-65-9 EINECS: 203-691-9 UN号: 1126 ADR: 3 II Merck Index: 14,01553 |
|
产品技术规格
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 98.0 % |