1,11-二溴十一烷 ≥96.0% (通过GC)
供应商: TCI
D2487-5G
D2487-1G
TCIAD2487-5GEA
0
CNY
TCIAD2487-5G
TCIAD2487-1G
1,11-二溴十一烷 ≥96.0% (通过GC)
1,11-二溴十一烷
分子式:
BrH₂C(CH₂)₉CH₂Br 沸点: 192°C 溶点: –11°C 密度: 1,332 g/cm³ (15 °C) 存储条件: Ambient |
MDL编号:
MFCD00000224 CAS号: 16696-65-4 EINECS: 240-745-0 |
|
产品技术规格
Appearance | Colorless to Orange to Green clear liquid |
Purity (GC) | min. 96.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...