(R)-(-)-仲丁胺 ≥98.0% (by GC, titration analysis)
供应商: TCI
|
Danger
|
Synonyms:
(R)-(-)-2-氨基丁烷
B2917-1G
B2917-100MG
TCIAB2917-1GEA
0
CNY
TCIAB2917-1G
TCIAB2917-100MG
(R)-(-)-仲丁胺 ≥98.0% (by GC, titration analysis)
(R)-(-)-仲丁胺
分子式:
C₄H₁₁N 沸点: 63°C 溶点: –104 °C 密度: 0,725 g/cm³ (20 °C) 闪点: -9°C 存储条件: Ambient |
MDL编号:
MFCD00064416 CAS号: 13250-12-9 EINECS: 236-232-6 UN号: 2733 ADR: 3,II Merck Index: 14,01544 |
|
产品技术规格
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 98.0 % |
Purity (Nonaqueous Titration) | min. 98.0 % |
Specific rotation [a]20/D | -6.0 to -10.0 deg (neat) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...