Cyclooctene ≥95.0% (by GC)

供应商: TCI
Warning

C0506-25ML C0506-500ML
TCIAC0506-25MLEA 0 CNY
TCIAC0506-25ML TCIAC0506-500ML
Cyclooctene ≥95.0% (by GC)
Cyclooctene
分子式: C₈H₁₄
分子量: 110.2 g/mol
沸点: 144°C
溶点: -16
闪点: 24°C
存储条件: Ambient
MDL编号: MFCD00001753
CAS号: 931-88-4
EINECS: 213-245-5
UN号: 3295
ADR: 3 III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=82825246c524f89a6d7dffdd53d829689a36da06&vid=64b431a5db2f05ae61af5218f5e508a12df34ef8&prid=web&clid=SAPDAM

现在订购

产品技术规格

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 95.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR