2-乙酰芴 ≥98.0% (通过GC)
供应商: TCI
A1069-25G
A1069-5G
TCIAA1069-25GEA
0
CNY
TCIAA1069-25G
TCIAA1069-5G
2-乙酰芴 ≥98.0% (通过GC)
2-乙酰芴
分子式:
C₁₅H₁₂O 沸点: 172 - 173 °C (0,05 -rr) 溶点: 128…132 °C |
MDL编号:
MFCD00001124 CAS号: 781-73-7 EINECS: 212-310-5 |
|
产品技术规格
Appearance | White to Light yellow powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 128.0 to 132.0 deg-C |
Solubility in hot Toluene | almost transparency |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...