2,7-二氨基芴 ≥98.0% (by HPLC)

供应商: TCI
Warning

Synonyms: 2,7-芴二胺

D0092-25G D0092-5G
TCIAD0092-25GEA 0 CNY
TCIAD0092-25G TCIAD0092-5G
2,7-二氨基芴 ≥98.0% (by HPLC)
2,7-二氨基芴
分子式: C₁₃H₁₂N₂
沸点: 467 °C (1013 hPa)
溶点: 165 °C
MDL编号: MFCD00001128
CAS号: 525-64-4
EINECS: 208-377-5
Merck Index: 14,04156
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c31e53ddda08b497ed11f607d9609b69b523b0bb&vid=c31e53ddda08b497ed11f607d9609b69b523b0bb&prid=web&clid=SAPDAM

现在订购

产品技术规格

Appearance White to Gray to Brown powder to crystaline
Purity (HPLC) min. 98.0 area%
Purity (Nonaqueous Titration) min. 97.0 %
Melting point 163.0 to 167.0 deg-C
Solubility in hot EtOH almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR