5′-溴-2′-羟基苯乙酮 ≥97.0% (by GC)

供应商: TCI
Warning

Synonyms: 1-(5-Bromo-2-hydroxyphenyl)ethan-1-one, 1-(5-Bromo-2-hydroxyphenyl)ethanone

B2061-25G B2061-5G
TCIAB2061-25GEA 0 CNY
TCIAB2061-25G TCIAB2061-5G
5′-溴-2′-羟基苯乙酮 ≥97.0% (by GC)
5′-溴-2′-羟基苯乙酮
分子式: C₈H₇BrO₂
分子量: 215.05
MDL编号: MFCD00191850
CAS号: 1450-75-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ec11a0bedfb714919cd6156c11eb40dc23d10505&vid=ec11a0bedfb714919cd6156c11eb40dc23d10505&prid=web&clid=SAPDAM

现在订购

产品技术规格

Appearance White to Yellow to Orange powder to crystal
Purity (GC) min. 97.0 %
Melting point 56.0 to 60.0 deg-C
Solubility in Methanol very faint turbidity


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR