L-薄荷醇 ≥99.0% (通过GC)

供应商: TCI
Warning

M0545-25G M0545-500G
TCIAM0545-25GEA 0 CNY
TCIAM0545-25G TCIAM0545-500G
L-薄荷醇 ≥99.0% (通过GC)
L-薄荷醇
分子式: C₁₀H₂₀O
沸点: 216°C
溶点: 41…44 °C
密度: 0,89 g/cm³ (20 °C)
闪点: 96 °C
存储条件: Ambient
MDL编号: MFCD00062979
CAS号: 2216-51-5
EINECS: 218-690-9
Merck Index: 14,05837
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7d56f87b67627ce9cdfb1a9a8b9e0b763008cccc&vid=7d56f87b67627ce9cdfb1a9a8b9e0b763008cccc&prid=web&clid=SAPDAM

现在订购


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR