1H-1,2,4-三氮唑-3-硫醇 ≥98.0% (by GC, titration analysis)
供应商: TCI
|
Warning
|
Synonyms:
3-巯基-1,2,4-三氮唑
M0814-25G
M0814-500G
TCIAM0814-25GEA
0
CNY
TCIAM0814-25G
TCIAM0814-500G
1H-1,2,4-三氮唑-3-硫醇 ≥98.0% (by GC, titration analysis)
1H-1,2,4-三氮唑-3-硫醇
分子式:
C₂H₃N₃S 分子量: 101.13 g/mol |
MDL编号:
MFCD00005229 CAS号: 3179-31-5 EINECS: 221-656-6 |
|
产品技术规格
Appearance | White to Light yellow powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Neutralization titration) | min. 98.0 % |
Melting point | 218.0 to 223.0 deg-C |
Solubility in hot Water | almost transparency |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...