原乙酸三乙酯 ≥96.0% (by GC)
供应商: TCI
|
Warning
|
Synonyms:
1,1,1-三乙氧基乙烷
O0105-25ML
O0105-500ML
TCIAO0105-25MLEA
0
CNY
TCIAO0105-25ML
TCIAO0105-500ML
原乙酸三乙酯 ≥96.0% (by GC)
原乙酸三乙酯
分子式:
C₈H₁₈O₃ 沸点: 145°C 密度: 0,8847 g/cm³ (20 °C) 闪点: 37°C |
MDL编号:
MFCD00009223 CAS号: 78-39-7 EINECS: 201-112-4 UN号: 3272 ADR: 3,III |
|
产品技术规格
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 96.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...