马尿酸 ≥98.0% (by titrimetric analysis)
供应商: TCI
|
Warning
|
Synonyms:
苯甲酰氨基乙酸
H0143-25G
H0143-500G
TCIAH0143-25GEA
0
CNY
TCIAH0143-25G
TCIAH0143-500G
马尿酸 ≥98.0% (by titrimetric analysis)
马尿酸
分子式:
C₆H₅C(O)NHCH₂COOH 分子量: 179.18 g/mol 沸点: 399 溶点: 271 |
MDL编号:
MFCD00002692 CAS号: 495-69-2 EINECS: 207-806-3 Merck Index: 14,04716 |
|
产品技术规格
Appearance | White powder to crystal |
Purity (Neutralization titration) | min. 98.0 % |
Melting point | 189.0 to 194.0 deg-C |
Solubility in hot Methanol | almost transparency |
Solubility in hot Water | almost transparency |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...