3,4-二乙氧基-3-环丁烯-1,2-二酮 ≥98.0% (通过GC)
供应商: TCI
|
Danger
|
D3253-25G
D3253-5G
TCIAD3253-25GEA
0
CNY
TCIAD3253-25G
TCIAD3253-5G
3,4-二乙氧基-3-环丁烯-1,2-二酮 ≥98.0% (通过GC)
3,4-二乙氧基-3-环丁烯-1,2-二酮
分子式:
C₈H₁₀O₄ 沸点: 88 - 92 °C (0,4 -rr) 密度: 1,15 g/cm³ (20 °C) |
MDL编号:
MFCD00001333 CAS号: 5231-87-8 |
|
产品技术规格
Appearance | Colorless to Orange to Green clear liquid |
Purity (GC) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...