2-甲氧基-1-萘醛 ≥98.0% (by GC)
供应商: TCI
M1040-25G
M1040-5G
TCIAM1040-25GEA
0
CNY
TCIAM1040-25G
TCIAM1040-5G
2-甲氧基-1-萘醛 ≥98.0% (by GC)
2-甲氧基-1-萘醛
分子式:
C₁₂H₁₀O₂ 分子量: 186.21 g/mol 沸点: 205°C |
MDL编号:
MFCD00004004 CAS号: 5392-12-1 EINECS: 226-392-5 |
|
产品技术规格
Appearance | White to Amber to Dark green powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 82.0 to 85.0 deg-C |
Solubility in hot Methanol | almost transparency |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...