乳酸丁酯 ≥98.0% (by GC)
供应商: TCI
|
Warning
|
Synonyms:
Butyl 2-hydroxypropanoate, n-Butyl lactate, Butyl (±)-2-hydroxypropionate, (±)-Butyl lactate
L0002-25G
L0002-500G
TCIAL0002-25GEA
0
CNY
TCIAL0002-25G
TCIAL0002-500G
乳酸丁酯 ≥98.0% (by GC)
乳酸丁酯
分子式:
C₇H₁₄O₃ 分子量: 146.19 g/mol 沸点: 187°C 溶点: -28 闪点: 91°C |
MDL编号:
MFCD00004519 CAS号: 138-22-7 EINECS: 205-316-4 |
|
产品技术规格
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 98.0 % |
Specific rotation [a]20/D | -10.0 to -13.0 deg (neat) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...