酚红钠盐

供应商: TCI
Warning

Synonyms: 酚磺酞

P0102-25G P0102-5G
TCIAP0102-25GEA 0 CNY
TCIAP0102-25G TCIAP0102-5G
酚红钠盐
酚红钠盐
分子式: C₁₉H₁₃NaO₅S
分子量: 376.35 g/mol
溶点: 285
存储条件: Ambient
MDL编号: MFCD00066901
CAS号: 34487-61-1
EINECS: 252-057-8
Merck Index: 12,07397
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4e5388e6dd5dae6c3503e03e409a592b374bf196&vid=4e5388e6dd5dae6c3503e03e409a592b374bf196&prid=web&clid=SAPDAM

现在订购

产品技术规格

Appearance Orange to Amber to Dark red powder to crystal
Absorbance min. 0.530 (5 mg/L, pH8.4, 556.0 to 560.0 nm)
Solubility in Water almost transparency
Drying loss max. 5.0 %
Transition interval (pH) (redish yellow)6.8 to 8.4 (red)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR