Alginic acid
供应商: TCI
A0733-500G
A0733-25G
TCIAA0733-500GEA
0
CNY
TCIAA0733-500G
TCIAA0733-25G
Alginic acid
Alginic acid
分子式:
(C₆H₈O₆)n 分子量: 176.13 g/mol 存储条件: Ambient |
MDL编号:
MFCD00081309 CAS号: 9005-32-7 EINECS: 232-680-1 Merck Index: 14,00242 |
产品技术规格
Appearance | White to Light yellow to Dark green powder to crystal |
Neutralization Value | 200.0 to 260.0 (calcd.on dried substance) |
Drying loss | max. 15.0 % |
Ignition residue (Sulfate) | max. 12.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...