3-噻吩甲腈 ≥98.0% (by GC)
供应商: TCI
|
Warning
|
Synonyms:
噻吩-3-甲腈, 3-氰基噻吩
C1344-25G
C1344-5G
TCIAC1344-25GEA
0
CNY
TCIAC1344-25G
TCIAC1344-5G
3-噻吩甲腈 ≥98.0% (by GC)
3-噻吩甲腈
分子式:
C₅H₃NS 分子量: 109.15 g/mol 沸点: 205°C |
MDL编号:
MFCD00151852 CAS号: 1641-09-4 EINECS: 216-687-7 |
|
产品技术规格
Appearance | Colorless to Light yellow clear liquid |
Purity (GC) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...