甲基红钾盐

供应商: TCI

Synonyms: 4-二甲氨基偶氮苯-2′-羧酸

M0424-1G M0424-25G
TCIAM0424-1GEA 0 CNY
TCIAM0424-1G TCIAM0424-25G
甲基红钾盐
甲基红钾盐
分子式: C₁₅H₁₄N₃NaO₂
分子量: 291.28 g/mol
存储条件: Ambient
MDL编号: MFCD00002426
CAS号: 845-10-3
EINECS: 212-682-9
Merck Index: 13,06142
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a57473fdbc215e9a7c363bd8faa2e592074db9d6&vid=a57473fdbc215e9a7c363bd8faa2e592074db9d6&prid=web&clid=SAPDAM

现在订购

产品技术规格

Appearance Orange to Brown powder to crystal
Transition interval (pH) (red)4.2 to 6.2 (yellow)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR