DL-丙氨酸 ≥98.5% (by titrimetric analysis)

供应商: TCI

Synonyms: (±)-2-氨基丙酸

A0178-25G A0178-500G
TCIAA0178-25GEA 0 CNY
TCIAA0178-25G TCIAA0178-500G
DL-丙氨酸 ≥98.5% (by titrimetric analysis)
DL-丙氨酸
分子式: C₃H₇NO₂
分子量: 89.09 g/mol
沸点: 189
溶点: 79
存储条件: Ambient
MDL编号: MFCD00064408
CAS号: 302-72-7
EINECS: 206-126-4
Merck Index: 14,00204
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=24aabc662b639c8a2dad52d43af1814cc1d8408c&vid=24aabc662b639c8a2dad52d43af1814cc1d8408c&prid=web&clid=SAPDAM

现在订购

产品技术规格

Appearance White powder to crystal
Purity (Nonaqueous Titration) min. 98.5 %
Solubility in Water almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR