2-溴-2-甲基丙酸 ≥98.0% (by GC)

供应商: TCI
Danger

Synonyms: 2-溴-2-甲基丙酸, α-溴异丁酸

B0605-25G B0605-500G
TCIAB0605-25GEA 0 CNY
TCIAB0605-25G TCIAB0605-500G
2-溴-2-甲基丙酸 ≥98.0% (by GC)
2-溴-2-甲基丙酸
分子式: C₄H₇BrO₂
分子量: 167 g/mol
溶点: 48
MDL编号: MFCD00004168
CAS号: 2052-01-9
EINECS: 218-139-2
UN号: 3261
ADR: 8 III
Merck Index: 14,01421
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=95a23b0c8daf88e442a920300e5d5154bd6c2657&vid=95a23b0c8daf88e442a920300e5d5154bd6c2657&prid=web&clid=SAPDAM

现在订购

产品技术规格

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 43.0 to 46.0 deg-C
Solubility in Water very faint turbidity


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR