(R)-(+)-氧化丙烯 ≥98.0% (by GC)

供应商: TCI
Danger

Synonyms: (R)-(+)-1,2-环氧丙烷, (R)-(+)-甲基环氧乙烷

P1396-25ML P1396-5ML
TCIAP1396-25MLEA 0 CNY
TCIAP1396-25ML TCIAP1396-5ML
(R)-(+)-氧化丙烯 ≥98.0% (by GC)
(R)-(+)-氧化丙烯
分子式: C₃H₆O
分子量: 58.08 g/mol
沸点: 34°C
闪点: –37°C
存储条件: Refrigerator
MDL编号: MFCD00066211
CAS号: 15448-47-2
UN号: 1280
ADR: 3 I
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=167f3c0d71e063b2cb9b1703520f5aab0781b769&vid=167f3c0d71e063b2cb9b1703520f5aab0781b769&prid=web&clid=SAPDAM

现在订购

产品技术规格

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %
Specific rotation [a]20/D +12.0 to +15.0 deg (neat)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR