己酸己酯 ≥98.0% (by GC)

供应商: TCI

Synonyms: Hexanoic acid hexyl ester

C1514-25ML C1514-500ML
TCIAC1514-25MLEA 0 CNY
TCIAC1514-25ML TCIAC1514-500ML
己酸己酯 ≥98.0% (by GC)
己酸己酯
分子式: C₁₂H₂₄O₂
沸点: 244°C
溶点: –55 °C
密度: 0,863 g/cm³ (25 °C)
MDL编号: MFCD00053808
CAS号: 6378-65-0
EINECS: 228-952-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0ed312b075ccc496d9b9f23030d880926edf41f3&vid=0ed312b075ccc496d9b9f23030d880926edf41f3&prid=web&clid=SAPDAM

现在订购

产品技术规格

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR