己酸丁酯 ≥98.0% (by GC)

供应商: TCI

Synonyms: Butyl caproate

H0107-25ML H0107-500ML
TCIAH0107-25MLEA 0 CNY
TCIAH0107-25ML TCIAH0107-500ML
己酸丁酯 ≥98.0% (by GC)
己酸丁酯
分子式: C₁₀H₂₀O₂
沸点: 208°C
密度: 0,866 g/cm³ (25 °C)
闪点: 102°C
MDL编号: MFCD00053804
CAS号: 626-82-4
EINECS: 210-964-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=191debaa3449aabf11ef86f6e4489fd4aba1284d&vid=191debaa3449aabf11ef86f6e4489fd4aba1284d&prid=web&clid=SAPDAM

现在订购

产品技术规格

Appearance Colorless clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR