异哌啶酸 ≥98.0% (by titrimetric analysis)

供应商: TCI
Warning

Synonyms: 六氢异烟酸, 4-哌啶甲酸

I0256-25G I0256-500G
TCIAI0256-25GEA 0 CNY
TCIAI0256-25G TCIAI0256-500G
异哌啶酸 ≥98.0% (by titrimetric analysis)
异哌啶酸
分子式: C₆H₁₁NO₂
分子量: 129.16 g/mol
沸点: 264
溶点: 157
MDL编号: MFCD00006004
CAS号: 498-94-2
EINECS: 207-872-3
Merck Index: 14,05189
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=bdbf5bc64a5f73f64dc73848fd00478c1e257379&vid=bdbf5bc64a5f73f64dc73848fd00478c1e257379&prid=web&clid=SAPDAM

现在订购

产品技术规格

Appearance White to Light yellow to Light red powder to crystal
Purity (Nonaqueous Titration) min. 98.0 %
Solubility in Water very faint turbidity


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR