3-哌啶甲酸 ≥98.0% (by titrimetric analysis)

供应商: TCI
Warning

Synonyms: 3-甲酸哌啶

N0420-25G N0420-500G
TCIAN0420-25GEA 0 CNY
TCIAN0420-25G TCIAN0420-500G
3-哌啶甲酸 ≥98.0% (by titrimetric analysis)
3-哌啶甲酸
分子式: C₆H₁₁NO₂
分子量: 129.16 g/mol
MDL编号: MFCD00005992
CAS号: 498-95-3
EINECS: 207-873-9
Merck Index: 14,06560
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3ee23314bbc50308e53175bf2fea70a33d6e9cc9&vid=3ee23314bbc50308e53175bf2fea70a33d6e9cc9&prid=web&clid=SAPDAM

现在订购

产品技术规格

Appearance White to Orange to Green powder to crystal
Purity (Nonaqueous Titration) min. 98.0 %
Solubility in Water very faint turbidity


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR