5,6-二氯烟酸 ≥98.0% (by titrimetric analysis)

供应商: TCI
Warning

Synonyms: 5,6-二氯烟酸, 5,6-二氯吡啶-3-羧酸

D2401-25G D2401-5G
TCIAD2401-25GEA 0 CNY
TCIAD2401-25G TCIAD2401-5G
5,6-二氯烟酸 ≥98.0% (by titrimetric analysis)
5,6-二氯烟酸
分子式: C₆H₃Cl₂NO₂
MDL编号: MFCD00075181
CAS号: 41667-95-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=25ca8a96d3751b831cef125dd37f943238649019&vid=25ca8a96d3751b831cef125dd37f943238649019&prid=web&clid=SAPDAM

现在订购

产品技术规格

Appearance White to Almost white powder to crystal
Purity (GC) min. 97.0 %
Purity (Neutralization titration) min. 98.0 %
Solubility in Methanol very faint turbidity


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR