2-溴吡啶-3-甲酸 ≥98.0% (by GC)
供应商: TCI
|
Warning
|
Synonyms:
2-溴烟酸
B2626-1G
B2626-5G
TCIAB2626-1GEA
0
CNY
TCIAB2626-1G
TCIAB2626-5G
2-溴吡啶-3-甲酸 ≥98.0% (by GC)
2-溴吡啶-3-甲酸
分子式:
C₆H₄BrNO₂ 分子量: 202.01 g/mol 溶点: 254°C |
MDL编号:
MFCD01646068 CAS号: 35905-85-2 |
|
产品技术规格
Appearance | White to Light yellow to Light orange powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Neutralization titration) | min. 97.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...