2,2,4-三甲基-1,3-戊二醇 ≥97.0% (by GC)

供应商: TCI
Warning

Synonyms: 2,2,4-Trimethyl-1,3-pentanediol, (±)-1,3-Dihydroxy-2,2,4-trimethyl pentane

T0483-25G T0483-500G
TCIAT0483-25GEA 0 CNY
TCIAT0483-25G TCIAT0483-500G
2,2,4-三甲基-1,3-戊二醇 ≥97.0% (by GC)
2,2,4-三甲基-1,3-戊二醇
分子式: C₈H₁₈O₂
沸点: 232°C
溶点: 51 °C
密度: 0,9485 g/cm³ (20 °C)
MDL编号: MFCD00004681
CAS号: 144-19-4
EINECS: 205-619-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=35dd4cfc50ef0d8e26a2ccdce2353da3dc13b729&vid=35dd4cfc50ef0d8e26a2ccdce2353da3dc13b729&prid=web&clid=SAPDAM

现在订购

产品技术规格

Appearance White to Almost white powder to crystal
Purity (GC) min. 97.0 %
Melting point 52.0 to 55.0 deg-C
Solubility in Methanol almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR