香芹酚 ≥98.0% (by GC)
供应商: TCI
Warning
Synonyms: 5-异丙基-2-甲基苯酚
C0026-25G C0026-500G
TCIAC0026-25GEA 0 CNY
TCIAC0026-25G TCIAC0026-500G
香芹酚 ≥98.0% (by GC)
香芹酚
分子式: C₁₀H₁₄O
分子量: 150.22 g/mol
沸点: 237
溶点: 0.5
闪点: 100°C
MDL编号: MFCD00002236
CAS号: 499-75-2
EINECS: 207-889-6
Merck Index: 14,01872
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e14352dca822e6f534da49e2ad2cbf1c3953e12c&vid=e14352dca822e6f534da49e2ad2cbf1c3953e12c&prid=web&clid=SAPDAM
现在订购
产品技术规格
Appearance Colorless to Light orange to Yellow clear liquid
Purity (GC) min. 98.0 %