N-羟乙基苯胺 ≥98.0% (by GC, titration analysis)
供应商: TCI
|
Danger
|
Synonyms:
2-苯胺乙醇, N-苯基乙醇胺, 2-苯氨基乙醇
A0466-25G
A0466-500G
TCIAA0466-25GEA
0
CNY
TCIAA0466-25G
TCIAA0466-500G
N-羟乙基苯胺 ≥98.0% (by GC, titration analysis)
N-羟乙基苯胺
分子式:
C₆H₅NHCH₂CH₂OH 分子量: 137.18 g/mol 沸点: 285 溶点: -30 闪点: 163°C |
MDL编号:
MFCD00002832 CAS号: 122-98-5 EINECS: 204-588-1 UN号: 2810 ADR: 6.1 II |
|
产品技术规格
Appearance | Colorless to Red to Green clear liquid |
Purity (GC) | min. 98.0 % |
Purity (Nonaqueous Titration) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...